Nickel(II) sulphate hexahydrate 98, GPR RECTAPUR®

Supplier: VWR Chemicals
Danger

Synonyms: Nickelous sulphate, Hexahydrate, Nickel sulphate hexahydrate

VWRC25895.294EA 116 SGD
VWRC25895.294
Nickel(II) sulphate hexahydrate 98, GPR RECTAPUR®
Nickel sulphate hexahydrate
Formula: NiSO₄·6H₂O
MW: 262.85 g/mol
Melting Pt: 840
Density: 2.07 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149813
CAS Number: 10101-97-0
EINECS: 232-104-9
UN: 3288
ADR: 6.1 III
Merck Index: 13,06541

Order Now

Specification Test Results

Assay Min. 98 %
Cl (Chloride) Max. 0.02 %
Cd (Cadmium) Max. 25 ppm
Cu (Copper) Max. 25 ppm
Pb (Lead) Max. 25 ppm
Zn (Zinc) Max. 25 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR