Magnesium chloride, anhydrous ≥98%, TECHNICAL

Supplier: VWR Chemicals

VWRC26123DCEA 1310 SGD
VWRC26123DC
Magnesium chloride, anhydrous ≥98%, TECHNICAL
Magnesium chloride
Formula: MgCl₂
Boiling Pt: 1412 °C (1013 hPa)
Melting Pt: 715 °C
Density: 2,32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011106
CAS Number: 7786-30-3
EINECS: 232-094-6
Merck Index: 13,05684

Order Now

Specification Test Results

Assay Min. 98 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR