Phenolphthalein, GPR RECTAPUR®

Supplier: VWR Chemicals
Danger

Synonyms: 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one

VWRC26237.231EA 133 SGD
VWRC26237.231
Phenolphthalein, GPR RECTAPUR®
Phenolphthalein
Formula: C₂₀H₁₄O₄
MW: 318.33 g/mol
Melting Pt: 258…262 °C
Density: 1.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005913
CAS Number: 77-09-8
EINECS: 201-004-7
REACH: 01-2119498295-24
Merck Index: 14,07243
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d81b539a32ba7c40a473e6afbb8e0dae525d637&vid=91bf141a7ea82baf5df563f73967884d2a4b5d79&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Suited for pH indicator Passes test
Melting point 259 - 264 °C
Ignition residue (SO4) Max. 0.1 %
Insolubility in ethanol 96 % (1% W/V) Max. 50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR