Lead(II) acetate trihydrate 99,5-103,0, AnalaR NORMAPUR® zA

Supplier: VWR Chemicals
Danger

Synonyms: Acetic acid lead (II) salt trihydrate, Lead diacetate trihydrate

VWRC26483.231EA 63.6 SGD
VWRC26483.231 VWRC26483.297
Lead(II) acetate trihydrate 99,5-103,0, AnalaR NORMAPUR® zA
Lead (II) acetate trihydrate
Formula: (H₃CCOO)₂Pb·3H₂O
MW: 379.34 g/mol
Boiling Pt: 280
Melting Pt: 75
Density: 2.55 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150023
CAS Number: 6080-56-4
EINECS: 206-104-4
UN: 1616
ADR: 6.1 III
Merck Index: 13,05415
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=351511a44fbb598a2d7fa1fa4ab395427b6168f1&vid=67e858e5f0dc4d04dd796541b3c0ad5aa73f8dc8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.5 - 103.0 %
pH (20°C; 10 %) 5.0 - 7.5
Insolubility in water Max. 50 ppm
Cl (Chloride) Max. 5 ppm
NO3 + NO2 (as NO3) Max. 100 ppm
Ca (Calcium) Max. 50 ppm
Cu (Copper) Max. 10 ppm
Fe (Iron) Max. 10 ppm
K (Potassium) Max. 50 ppm
Na (Sodium) Max. 50 ppm
Conforms to Reag. Ph.Eur. Passes test
Conforms to ACS Passes test

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR