Potassium bromide ≥98.5%, GPR RECTAPUR®

Supplier: VWR Chemicals
Warning

VWRC26704.292EA 153 SGD
VWRC26704.292
Potassium bromide ≥98.5%, GPR RECTAPUR®
Potassium bromide
Formula: KBr
MW: 119 g/mol
Boiling Pt: 1435
Melting Pt: 730
Density: 2.75 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011358
CAS Number: 7758-02-3
EINECS: 231-830-3
Merck Index: 13,07701

Order Now

Specification Test Results

Assay Min. 98.5 %
Heavy metals (as Pb) Max. 10 ppm
Cl (Chloride) Max. 0.5 %
SO4 (Sulphate) Max. 100 ppm
Fe (Iron) Max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR