Ammonium cerium(IV) sulphate 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution

Supplier: VWR Chemicals
Warning

Synonyms: Tetraammonium cerium(IV) sulphate, Cerium (IV) ammonium sulphate, Ammonium ceric sulphate, Ammonium cerium sulphate hydrate, Ammonium cerium(IV) sulphate hydrate, Tetraammonium cerium tetrakis(sulphate)

VWRC2678.1000EA 607 SGD
VWRC2678.1000
Ammonium cerium(IV) sulphate 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution
Ammonium cerium (IV) sulphate
Formula: H₁₆CeN₄O₁₆S₄
MW: 596.52
MDL Number: MFCD00148851
CAS Number: 7637-03-8
EINECS: 231-567-4
UN: 3264
ADR: 8 III

Order Now

Specification Test Results

Titer (20°C; real value 0.2 % accuracy) 0.0998 - 0.1002 mol/l
NIST traceable Confirmed


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR