Potassium permanganate ≥99%, GPR RECTAPUR®

Supplier: VWR Chemicals
Danger

Synonyms: Permanganic acid potassium salt

VWRC26906.290EA 68.4 SGD
VWRC26906.290 VWRC26906.368
Potassium permanganate ≥99%, GPR RECTAPUR®
Potassium permanganate
Formula: KMnO₄
MW: 158.03 g/mol
Melting Pt: > 240 °C
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011364
CAS Number: 7722-64-7
EINECS: 231-760-3
UN: 1490
ADR: 5.1 II
Merck Index: 12,07824

Order Now

Specification Test Results

Assay Min. 99 %
Cl (Chloride) Max. 0.02 %
SO4 (Sulphate) Max. 0.02 %
Pb (Lead) Max. 50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR