Potassium permanganate 99,0-100,5, AnalaR NORMAPUR® Reag. Ph. Eur. zA

Supplier: VWR Chemicals
Danger

Synonyms: Permanganic acid potassium salt

VWRC26910.410EA 607 SGD
VWRC26910.410
Potassium permanganate 99,0-100,5, AnalaR NORMAPUR® Reag. Ph. Eur. zA
Potassium permanganate
Formula: KMnO₄
MW: 158.03 g/mol
Melting Pt: > 240 °C
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011364
CAS Number: 7722-64-7
EINECS: 231-760-3
UN: 1490
ADR: 5.1 II
Merck Index: 12,07824

Order Now

Specification Test Results

Assay 99.0 - 100.5 %
Appearance of solution Passes test Ph.Eur.
Identification A Passes test Ph.Eur.
Identification B Passes test Ph.Eur.
Insolubility in water Max. 0.1 %
Total N (Nitrogen) Max. 50 ppm
Cl (Chloride) Max. 0.02 %
SO4 (Sulphate) Max. 100 ppm
Fe (Iron) Max. 20 ppm
Pb (Lead) Max. 20 ppm
Conforms to Reag. Ph.Eur. Passes test

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR