Potassium sulphate ≥99%, GPR RECTAPUR®

Supplier: VWR Chemicals

VWRC26994.293EA 51.3 SGD
VWRC26994.293 VWRC26994.362 VWRC26994.460
Potassium sulphate ≥99%, GPR RECTAPUR®
Potassium sulphate
Formula: K₂SO₄
MW: 174.26 g/mol
Boiling Pt: 1698
Melting Pt: 1069 °C
Density: 2.66 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011388
CAS Number: 7778-80-5
EINECS: 231-915-5
Merck Index: 13,07759

Order Now

Specification Test Results

Assay Min. 99 %
Heavy metals (as Pb) Max. 20 ppm
Cl (Chloride) Max. 50 ppm
NH4 (Ammonium) Max. 20 ppm
NO3 (Nitrate) Max. 50 ppm
Fe (Iron) Max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR