Peqlab peqGOLD, Protein Marker II (Unstained)
Supplier: VWR Chemicals
VWRC27-2010EA 385 SGD
VWRC27-2010
Peqlab peqGOLD, Protein Marker II (Unstained)
Electrophoresis Reagents Molecular Weight Markers
Protein Marker II for accurate molecular weight determination of proteins by SDS-PAGE.
  • Mix of 14 recombinant proteins
  • Range: 10 to 200 kDa
  • Each protein with Strep-Tag® II sequence (WSHPQFEK)
  • Highlighted 50 kDa band for easy orientation
  • Ready to use for a fast, convenient workflow