Potassium hydrogen sulphate 99,0, AnalaR NORMAPUR® zA

Supplier: VWR Chemicals
Danger

Synonyms: Potassium bisulphate

VWRC27011.237EA 25.9 SGD
VWRC27011.237 VWRC27011.294 VWRC27011.460
Potassium hydrogen sulphate 99,0, AnalaR NORMAPUR® zA
Potassium hydrogen sulphate
Formula: KHSO₄
MW: 136.17 g/mol
Melting Pt: 214
Density: 2.32 g/cm³ (20 °C)
MDL Number: MFCD00011404
CAS Number: 7646-93-7
EINECS: 231-594-1
UN: 2509
ADR: 8 II
Merck Index: 13,07695

Order Now

Specification Test Results

Assay Min. 99.0 %
Heavy metals (as Pb) Max. 5 ppm
Total N (Nitrogen) Max. 10 ppm
Cl (Chloride) Max. 5 ppm
PO4 (Phosphate) Max. 10 ppm
Al (Aluminium) Max. 20 ppm
Ca (Calcium) Max. 50 ppm
Fe (Iron) Max. 5 ppm
Na (Sodium) Max. 50 ppm

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR