Benzyl alcohol 97-100,5, GPR RECTAPUR®

Supplier: VWR Chemicals
Warning

Synonyms: Benzenemethanol, Phenylmethanol, Phenylcarbinol

VWRC27354EEEA 1320 SGD
VWRC27354EE VWRC27354UU
Benzyl alcohol 97-100,5, GPR RECTAPUR®
Benzyl alcohol
Formula: C₆H₅CH₂OH
MW: 108.14 g/mol
Boiling Pt: 205
Melting Pt: –15,3 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 101
Storage Temperature: Ambient
MDL Number: MFCD00004599
CAS Number: 100-51-6
EINECS: 202-859-9
Merck Index: 13,01126
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0c85c958ac64f8b86c5836e49f2c4354b22886d&vid=633ad635461465914e782d5a22480c9ff8f81bf7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 97 - 100.5 %
Acidity Passes test
Appearance of solution Passes test
IR Spectrum Passes test
Density (20/20) 1.043 - 1.049
Peroxide value Max. 5
Refractive index (20°C) 1.538 - 1.541
Benzaldehyde Max. 0.15 %
Evaporation residue Max. 0.05 %
Related substances Max. 0.2 %
Cl (Chloride) Max. 20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR