Sand Fontainebleau, TECHNICAL

Supplier: VWR Chemicals

VWRC27460.295EA 34.9 SGD
VWRC27460.295 VWRC27460.364 VWRC27460.460
Sand Fontainebleau, TECHNICAL
Sand Fontainebleau
Formula: SiO₂
MW: 60.08 g/mol
Boiling Pt: 2230 °C (1013 hPa)
Storage Temperature: Ambient
CAS Number: 7631-86-9
EINECS: 238-878-4

Order Now

Specification Test Results

Identification Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR