Sand, TECHNICAL, industrial, washed

Supplier: VWR Chemicals

Synonyms: Silicium dioxide, Silicium (IV) oxide, Silicon dioxide

VWRC27461.298EA 32.7 SGD
VWRC27461.298 VWRC27461.360 VWRC27461.460
Sand, TECHNICAL, industrial, washed
Sand
Formula: SiO₂
MW: 60.08 g/mol
Melting Pt: 1710 °C
Density: 2.66 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD02100519
CAS Number: 14808-60-7
EINECS: 238-878-4

Order Now

Specification Test Results

Solubility in acid Max. 1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR