Sodium alginate, TECHNICAL

Supplier: VWR Chemicals

Synonyms: Alginic acid sodium salt, Algin

VWRC27660.296EA 154 SGD
VWRC27660.296
Sodium alginate, TECHNICAL
Sodium alginate
Storage Temperature: Ambient
MDL Number: MFCD00081310
CAS Number: 9005-38-3

Order Now

Specification Test Results

Identification Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR