Sodium carbonate ≥97%, TECHNICAL

Supplier: VWR Chemicals
Warning

VWRC27754.360EA 88.8 SGD
VWRC27754.360 VWRC27754.460
Sodium carbonate ≥97%, TECHNICAL
Sodium carbonate
Formula: Na₂CO₃
Boiling Pt: 1600 °C (1013 hPa)
Melting Pt: 854 °C
Density: 2,53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003494
CAS Number: 497-19-8
EINECS: 207-838-8
REACH: 01-2119485498-19
Merck Index: 14,08596
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a6ff30f1487cdef86add98b38bfa8eb3dac14f6&vid=c5b129b7acb5c897b9d970ea3130d61fb1934b54&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (on anhydrous substance) Min. 97 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR