Sodium carbonate ≥99%, GPR RECTAPUR®

Supplier: VWR Chemicals
Warning

Synonyms: Calcined soda, Carbonic acid disodium salt

VWRC27766.292EA 65.4 SGD
VWRC27766.292 VWRC27766.361 VWRC27766.463
Sodium carbonate ≥99%, GPR RECTAPUR®
Sodium carbonate
Formula: Na₂CO₃
MW: 105.99 g/mol
Boiling Pt: 1600
Melting Pt: 854
Density: 2.53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003494
CAS Number: 497-19-8
EINECS: 207-838-8
REACH: 01-2119485498-19
Merck Index: 14,08596
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a6ff30f1487cdef86add98b38bfa8eb3dac14f6&vid=c5b129b7acb5c897b9d970ea3130d61fb1934b54&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (on anhydrous substance) Min. 99 %
Heavy metals (as Pb) Max. 20 ppm
Cl (Chloride) Max. 100 ppm
SO4 (Sulphate) Max. 100 ppm
Fe (Iron) Max. 20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR