Sodium carbonate, anhydrous 99.5-100.5% Ph. Eur., NF, BP

Supplier: VWR Chemicals
Warning

VWRC27767.295EA 57.5 SGD
VWRC27767.295 VWRC27767.466
Sodium carbonate, anhydrous 99.5-100.5% Ph. Eur., NF, BP
Sodium carbonate

Appearance: White or almost white, slightly granular powder, hygroscopic.


Caution: Not for use as Active Pharmaceutical Ingredient (API)

Formula: Na₂CO₃
Boiling Pt: 1600 °C (1013 hPa)
Melting Pt: 854 °C
Density: 2,53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003494
CAS Number: 497-19-8
EINECS: 207-838-8
REACH: 01-2119485498-19
Merck Index: 14,08596
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a6ff30f1487cdef86add98b38bfa8eb3dac14f6&vid=c5b129b7acb5c897b9d970ea3130d61fb1934b54&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Conforms (see CoA/CoS)
Heavy metals (as Pb) ≤ 50 ppm
Residual solvents Unlikely by manuf.process
Residues of metal catalysts or reagents Unlikely by manuf.process
As (Arsenic) ≤ 5 ppm
Cd (Cadmium) ≤ 1 ppm
Co (Cobalt) ≤ 1 ppm
Hg (Mercury) ≤ 2 ppm
Ni (Nickel) ≤ 1 ppm
Pb (Lead) ≤ 5 ppm
V (Vanadium) ≤ 6 ppm
Conforms to Ph.Eur./NF/BP Passes test
Ph.Eur./BP specifications: .
Assay (dried substance) [Ph.Eur./BP] 99.5 - 100.5 %
Identification A Passes test NF
Identification B Passes test NF
Identification C Passes test Ph.Eur./BP
Identification D [Ph.Eur./BP] ≤ 1.0 %
Appearance of solution Passes test Ph.Eur./BP
Alkali hydroxides and bicarbonates Passes test Ph.Eur./BP
Cl (Chloride) [Ph.Eur./BP] ≤ 125 ppm
SO4 (Sulphate) [Ph.Eur./BP] ≤ 250 ppm
Fe (Iron) [Ph.Eur./BP] ≤ 50 ppm
Loss on drying (300 ± 15°C) ≤ 1.0 %
NF specifications: .
Assay (calculated on anhydrous) [NF] 99.5 - 100.5 %
Water determination [NF] ≤ 0.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR