Sodium gluconate, TECHNICAL

Supplier: VWR Chemicals

Synonyms: D-Gluconic acid sodium salt, Sodium (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate, Gluconic acid, sodium salt, Sodium D-gluconate

VWRC27871.295EA 54.2 SGD
VWRC27871.295 VWRC27871.460
Sodium gluconate, TECHNICAL
Sodium gluconate
Formula: HOH₂C(CH(OH))₄COONa
Melting Pt: 170…175 °C
Storage Temperature: Ambient
MDL Number: MFCD00064210
CAS Number: 527-07-1
EINECS: 208-407-7
Merck Index: 12,08766
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eafd5b786cb23fb5c20dd64ad056ed2d80dc0186&vid=b1ffe48dafca4b8bd9b0d709ea3e986099051767&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identification Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR