DL-Sodium lactate 60% in aqueous solution, purified

Supplier: VWR Chemicals

Synonyms: (±)-Lactic acid sodium salt, (±)-2-Hydroxypropionic acid sodium salt, Lactate sodium, DL-Lactic acid sodium salt, (±)-2-Hydroxypropanoic acid sodium salt, Sodium 2-hydroxypropanoate

VWRC27925.292EA 155 SGD
VWRC27925.292
DL-Sodium lactate 60% in aqueous solution, purified
DL-Sodium lactate
Formula: H₃CCH(OH)COONa
MW: 112.06 g/mol
Density: 1.27…1.32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00065400
CAS Number: 72-17-3
EINECS: 200-772-0
Merck Index: 13,08709

Order Now

Specification Test Results

Assay 59,0-61,5 %
Heavy metals (as Pb) Max. 10 ppm
Fe (Iron) Max. 5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR