DL-Sodium lactate 50% in aqueous solution Ph. Eur.

Supplier: VWR Chemicals

VWRC27927.298EA 97.9 SGD
VWRC27927.298 VWRC27927.367
DL-Sodium lactate 50% in aqueous solution Ph. Eur.
DL-Sodium lactate
Formula: H₃CCH(OH)COONa
Density: 1,263 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00065400
CAS Number: 72-17-3
EINECS: 200-772-0

Order Now

Specification Test Results

Assay (Sodium 2-hydroxypropanoate) Min. 50%
Appearance Conforms (see CoA/CoS)
Identification A Passes test
Identification B Passes test
Appearance of solution Passes test
pH 6.5 - 9.0
Reducing sugars and sucrose Passes test
Chlorides (calc. ref. to Na lactate) Max. 50 ppm
Oxalates and phosphates Passes test
Sulphates (calc. ref. to Na lactate) Max. 100 ppm
Iron (calc. ref. to Na lactate) Max. 10 ppm
Heavy metals (calc. ref. to Na lactate) Max. 10 ppm
Residual solvents Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR