Sodium periodate ≥99.0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent

Supplier: VWR Chemicals
Danger

Synonyms: Sodium metaperiodate

VWRC27985.185EA 63 SGD
VWRC27985.185 VWRC27985.290 VWRC27985.230
Sodium periodate ≥99.0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Sodium periodate
Formula: NaIO₄
MW: 213.89 g/mol
Boiling Pt: 300
Melting Pt: 175
Density: 3.87 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003534
CAS Number: 7790-28-5
EINECS: 232-197-6
UN: 1479
ADR: 5.1 II
Merck Index: 13,08714

Order Now

Specification Test Results

Assay Min. 99.0 %
Assay (dried basis) 99.8-100.3 %
Cl + ClO3 + Br + BrO3 (as Cl) Max. 100 ppm
SO4 (Sulphate) Max. 50 ppm
Mn (Manganese) Max. 1 ppm
Conforms to ACS Passes test
Conforms to Reag. Ph.Eur. Passes test

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR