Sodium salicylate ≥99.5%, AnalaR NORMAPUR® analytical reagent

Supplier: VWR Chemicals
Warning

Synonyms: Sodium 2-hydroxybenzoate, 2-Hydroxybenzoic acid sodium salt, Salicylic acid sodium salt

VWRC28065.230EA 49.5 SGD
VWRC28065.230
Sodium salicylate ≥99.5%, AnalaR NORMAPUR® analytical reagent
Sodium salicylate
Formula: HOC₆H₄COONa
Melting Pt: 200 °C
Density: 0,32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002440
CAS Number: 54-21-7
EINECS: 200-198-0
Merck Index: 12,08819

Order Now

Specification Test Results

Assay (calculated on dried substance) Min. 99.5 %
Heavy metals (as Pb) Max. 10 ppm
Water Max. 0.2 %
Cl (Chloride) Max. 20 ppm
SO4 (Sulphate) Max. 0.02 %
Fe (Iron) Max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR