Vanadyl sulphate pentahydrate, TECHNICAL

Supplier: VWR Chemicals
Warning

VWRC28900.180EA 103 SGD
VWRC28900.180
Vanadyl sulphate pentahydrate, TECHNICAL
Vanadyl sulphate pentahydrate
Formula: VOSO₄·5H₂O
Density: 2 g/cm³ (20 °C)
MDL Number: MFCD00149933
CAS Number: 12439-96-2
EINECS: 248-652-7
UN: 2931
ADR: 6.1,II

Order Now

Specification Test Results

Identification Passes test

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR