D-(+)-Xylose, GPR RECTAPUR®

Supplier: VWR Chemicals

VWRC29013.237EA 60.6 SGD
VWRC29013.237
D-(+)-Xylose, GPR RECTAPUR®
D(+)-Xylose
Formula: C₅H₁₀O₅
MW: 150.13 g/mol
Boiling Pt: 331
Melting Pt: 154
Density: 1.525 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00151475
CAS Number: 58-86-6
EINECS: 200-400-7
Merck Index: 13,10142
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f045c9c134fcd7d287b95738095c40d429a173e&vid=447bd80bb2f3ca1cb52dc711d0957e9ab6e2f84d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Melting point 145 - 150 °C
Specific optical rotation (10 %; water) 18 - 20 °
Ignition residue (SO4) Max. 0.2 %
Loss on drying (100°C) Max. 0.2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR