Magnesium sulphate, (dried basis) ≥98%, GPR RECTAPUR®

Supplier: VWR Chemicals

Synonyms: MgSO4, Magnesium sulphate anhydrous

VWRC291184PEA 25.9 SGD
VWRC291184P VWRC291186R VWRC29118BW
Magnesium sulphate, (dried basis) ≥98%, GPR RECTAPUR®
Magnesium sulphate
Formula: MgSO₄
MW: 120.37 g/mol
Melting Pt: 1127 °C
Density: 2.65 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011110
CAS Number: 7487-88-9
EINECS: 231-298-2
Merck Index: 12,05731
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bc2ae189b0cbc1bfc599951528466503d0d9008&vid=63867221020ebd2c8c227492e609a4764639ff79&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 98 %
Heavy metals (as Pb) Max. 10 ppm
Cl (Chloride) Max. 0.05 %
Fe (Iron) Max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR