Periodic acid 50%, GPR RECTAPUR®

Supplier: VWR Chemicals
Danger

Synonyms: ortho-Periodic acid, Orthoperiodic acid

VWRC294604DEA 124 SGD
VWRC294604D
Periodic acid 50%, GPR RECTAPUR®
Periodic acid
Formula: H₅IO₆
MW: 227.94 g/mol
Melting Pt: 122
MDL Number: MFCD00011344
CAS Number: 10450-60-9
EINECS: 233-937-0
UN: 3098
ADR: 5.1 (8) II
Merck Index: 13,07248
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39e1882cdf619581f467ea2429265a700b5205d5&vid=27e789fa0f30c08d62d5a8d0afc3acd8f6469c12&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 48 - 53 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR