Polyvinyl alcohol, M.W. 115,000 ≥88%, hydrolysed

Supplier: VWR Chemicals

Synonyms: Poly(vinyl alcohol), PVOH, PVA, Mowiol®

VWRC297914DEA 146 SGD
VWRC297914D
Polyvinyl alcohol, M.W. 115,000 ≥88%, hydrolysed
Polyvinyl alcohol
Formula: [-CH₂CH(OH)-]n
MDL Number: MFCD00081922
CAS Number: 9002-89-5

Order Now

Specification Test Results

Appearance White granular powder
pH (4 %) 4.5 - 6.5
Viscosity (20°C; 4 %; water) 46 - 54 mPa.s
Degree of hydrolysis 86.5 - 89 %
Ignition residue Max. 0.5 %
Loss on drying (60°C) Max. 5 %
Organic volatile impurities Max. 1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR