Oxalic acid 0.5 mol/l (1 N) in aqueous solution, AVS TITRINORM volumetric solution

Supplier: VWR Chemicals

Synonyms: Ethanedioic acid

VWRC30086.293EA 55.4 SGD
VWRC30086.293
Oxalic acid 0.5 mol/l (1 N) in aqueous solution, AVS TITRINORM volumetric solution
Oxalic acid
Formula: C₂H₂O₄
Storage Temperature: Ambient
MDL Number: MFCD00002573
CAS Number: 144-62-7
EINECS: 205-634-3
ADR: 8,III

Order Now

Specification Test Results

Titer (20°C; real value 0.2 % accuracy) 0.499 - 0.501 mol/l
NIST traceable Confirmed


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR