Oxalic acid 0.05 mol/l (0.1 N), AVS TITRINORM volumetric solution

Supplier: VWR Chemicals

Synonyms: Ethanedioic acid

VWRC30093.297EA 41.8 SGD
VWRC30093.297
Oxalic acid 0.05 mol/l (0.1 N), AVS TITRINORM volumetric solution
Oxalic acid
Formula: C₂H₂O₄
Storage Temperature: Ambient
MDL Number: MFCD00002573
CAS Number: 144-62-7
EINECS: 205-634-3
ADR: 8,III

Order Now

Specification Test Results

Titer (20°C; real value 0.2 % accuracy) 0.0499 - 0.0501 mol/l
NIST traceable Confirmed


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR