Paraffin, liquid, sterile
Supplier: VWR Chemicals
Synonyms: Wax Paraffin, Paraffin wax
VWRC301440ZKEA 57.2 SGD
VWRC301440ZK
Paraffin, liquid, sterile
Paraffin
Sterile liquid paraffin is mostly used in microbiology as a vehicle to improve the anaerobic culture media as: O / F, decarboxylase medium, MIO, etc. Typically the product (approx 2 ml) is aseptically dispensed on the surface of the sterile culture medium in a culture tube.
  • Supplied in a tightly closed container with good seal, protected from light
  • Colourless
Raw material complies with DAB10, USP XXI Eur. Pharm and B.P.
CAS Number: 8012-95-1