Zinc sulphate 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution

Supplier: VWR Chemicals

VWRC30498.296EA 46.8 SGD
VWRC30498.296
Zinc sulphate 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution
Zinc sulphate
Formula: O₄SZn
MW: 161.45 g/mol
Melting Pt: 100
Density: 1.45
Storage Temperature: Ambient
MDL Number: MFCD00011302
CAS Number: 7733-02-0
EINECS: 231-793-3
UN: 3082
ADR: 9 III
Merck Index: 12,10293

Order Now

Specification Test Results

NIST traceable Confirmed
Titer (20°C; real value 0.2 % accuracy) 0.0998 - 0.1002 mol/l


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR