Polyvinyl alcohol, M.W. 22,000 ≥98%, hydrolysed

Supplier: VWR Chemicals

VWRC305735BEA 96.5 SGD
VWRC305735B
Polyvinyl alcohol, M.W. 22,000 ≥98%, hydrolysed
Polyvinyl alcohol
Formula: [-CH₂CH(OH)-]n
MDL Number: MFCD00081922
CAS Number: 9002-89-5

Order Now

Specification Test Results

Viscosity (20°C; 4 %; water) 5,5 - 6,6 mPa.s
Degree of hydrolysis Min. 98 %
Ignition residue (SO4) Max. 1 %
Loss on drying (105°C) Max. 5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR