Barium chloride 1 g/l, VOLUSOL® for hydrometry

Supplier: VWR Chemicals

VWRC30594.298EA 88.5 SGD
VWRC30594.298
Barium chloride 1 g/l, VOLUSOL® for hydrometry
Barium chloride
Formula: BaCl₂
Storage Temperature: Ambient
MDL Number: MFCD00003445
CAS Number: 10361-37-2
EINECS: 241-234-5
UN: 3287
ADR: 6.1,III

Order Now

Specification Test Results

Assay (complexometric, calc. on the ignition residue) Min 99 %
Loss on ignition (1000 °C) Max 1.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR