Ammonium iron(II) sulphate 0.05 mol/l (0.05 N) in aqueous solution

Supplier: VWR Chemicals

Synonyms: di-Ammonium iron (II) sulphate, Ammonium ferrous sulphate

VWRC30882.294EA 0 SGD
VWRC30882.294
Ammonium iron(II) sulphate 0.05 mol/l (0.05 N) in aqueous solution
Ammonium iron (II) sulphate
Formula: H₈FeN₂O₈S₂
MW: 284.05 g/mol
Melting Pt: 100
Density: 1.86
Storage Temperature: Ambient
MDL Number: MFCD00051039
CAS Number: 10045-89-3
EINECS: 233-151-8
Merck Index: 12,00552

Order Now

Specification Test Results

Titer (20°C; real value 0.2 % accuracy) 0.0499 - 0.0501 mol/l
NIST traceable Confirmed


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR