Ammonium iron(II) sulphate 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution

Supplier: VWR Chemicals

Synonyms: di-Ammonium iron (II) sulphate, Ammonium ferrous sulphate

VWRC30883.297EA 75.7 SGD
VWRC30883.297
Ammonium iron(II) sulphate 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution
Ammonium iron (II) sulphate

Not suitable for extended storage. Check strength before use.

Formula: H₈FeN₂O₈S₂
MW: 284.05 g/mol
Melting Pt: 100
Density: 1.030 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00051039
CAS Number: 10045-89-3
EINECS: 233-151-8
Merck Index: 12,00552

Order Now

Specification Test Results

Titer (20°C, at filling) 0.0998 - 0.1002 mol/l
NIST traceable Confirmed
Measure the titer before use.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR