Potassium dichromate 0.04 mol/l (0.24 N) with mercury (II) sulphate 80 g/l in sulphuric acid solution, AVS TITRINORM volumetric solution, for COD determination in water

Supplier: VWR Chemicals
Danger

VWRC30891.295EA 198 SGD
VWRC30891.295 VWRC30891.420
Potassium dichromate 0.04 mol/l (0.24 N) with mercury (II) sulphate 80 g/l in sulphuric acid solution, AVS TITRINORM volumetric solution, for COD determination in water
Potassium dichromate

According to NFT 90-101 standard

Formula: K₂Cr₂O₇
Boiling Pt: >100 °C (1013 hPa)
Density: 1,19 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011367
CAS Number: 7778-50-9
EINECS: 231-906-6
UN: 3289
ADR: 6.1 (8),II

Order Now

Specification Test Results

Titer (20°C; real value 0.2 % accuracy) 0.03992 - 0.04008 mol/l
NIST traceable Confirmed


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR