Nitric acid 2 mol/l (2 N), AVS TITRINORM volumetric solution

Supplier: VWR Chemicals
Danger

Synonyms: Nitric acid solution

VWRC310568.1000EA 75.7 SGD
VWRC310568.1000
Nitric acid 2 mol/l (2 N), AVS TITRINORM volumetric solution
Nitric acid
Formula: HNO₃
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
UN: 2031
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR