Potassium permanganate 0.2 mol/l (1 N) in aqueous solution, AVS TITRINORM volumetric solution

Supplier: VWR Chemicals

Synonyms: Permanganic acid potassium salt

VWRC31451.292EA 79 SGD
VWRC31451.292 VWRC31451.326
Potassium permanganate 0.2 mol/l (1 N) in aqueous solution, AVS TITRINORM volumetric solution
Potassium permanganate
Formula: KMnO₄
MW: 158.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011364
CAS Number: 7722-64-7
EINECS: 231-760-3
UN: 3082
ADR: 9 III
Merck Index: 12,07824

Order Now

Specification Test Results

Titer (20°C; real value 0.2 % accuracy) 0.1996 - 0.2004 mol/l
NIST traceable Confirmed

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR