Potassium permanganate 0.02 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution

Supplier: VWR Chemicals

VWRC31455.295EA 37.2 SGD
VWRC31455.295 VWRC31455.320
Potassium permanganate 0.02 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution
Potassium permanganate
Formula: KMnO₄
Storage Temperature: Ambient
MDL Number: MFCD00011364
CAS Number: 7722-64-7
EINECS: 231-760-3
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Titer (20°C; real value 0.2 % accuracy) 0.01996 - 0.02004 mol/l
NIST traceable Confirmed


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR