Potassium permanganate 0,02 mol/l 0,1 N in aqueous solution, AVS TITRINORM volumetric solution standardised against ferrous ethylenediammonium sulphate

Supplier: VWR Chemicals

Synonyms: Permanganic acid potassium salt

VWRC31456.295EA 57.6 SGD
VWRC31456.295
Potassium permanganate 0,02 mol/l 0,1 N in aqueous solution, AVS TITRINORM volumetric solution standardised against ferrous ethylenediammonium sulphate
Potassium permanganate
Formula: KMnO₄
MW: 158.03 g/mol
MDL Number: MFCD00011364
CAS Number: 7722-64-7
UN: 3082
ADR: 9 III
Merck Index: 12,07824

Order Now

Specification Test Results

Titer (20°C; real value 0.2% accuracy) 0.01996 - 0.02004 mol/L
NIST traceable Confirmed
Conforms to Reag.Ph.Eur. Passes test
Standardised against C₂H₈N₂.Fe.2H₂SO₄ Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR