Sodium carbonate 0.5 mol/l (1 N) aqueous solution, AVS TITRINORM Volumetric

Supplier: VWR Chemicals
Warning

Synonyms: Calcined soda, Carbonic acid disodium salt

VWRC31524.294EA 53.3 SGD
VWRC31524.294
Sodium carbonate 0.5 mol/l (1 N) aqueous solution, AVS TITRINORM Volumetric
Sodium carbonate
Formula: Na₂CO₃
MW: 105.99 g/mol
Boiling Pt: 1600
Melting Pt: 854
Storage Temperature: Ambient
MDL Number: MFCD00003494
CAS Number: 497-19-8
EINECS: 207-838-8
Merck Index: 14,08596
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a6ff30f1487cdef86add98b38bfa8eb3dac14f6&vid=c5b129b7acb5c897b9d970ea3130d61fb1934b54&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Titer (20°C; real value 0.2 % accuracy) 0.499 - 0.501 mol/l
NIST traceable Confirmed

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR