Sodium chloride 1 mol/l (1 N) in aqueous solution, AVS TITRINORM volumetric solution

Supplier: VWR Chemicals

Synonyms: Saline, NaCl, Sea salt, Halite

VWRC31535.292EA 75.8 SGD
VWRC31535.292
Sodium chloride 1 mol/l (1 N) in aqueous solution, AVS TITRINORM volumetric solution
Sodium chloride
Formula: NaCl
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3

Order Now

Specification Test Results

Titer (20°C; real value 0.2 % accuracy) 0.998 - 1.002 mol/l
NIST traceable Confirmed


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR