Phenolphthalein 0,2% in ethanolic solution, TECHNICAL

Supplier: VWR Chemicals
Danger

VWRC31724.295EA 71 SGD
VWRC31724.295
Phenolphthalein 0,2% in ethanolic solution, TECHNICAL
Phenolphthalein

  • Transition range: pH 8,2-9,8

Formula: C₂₀H₁₄O₄
Density: 0,884 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005913
CAS Number: 77-09-8
EINECS: 201-004-7
UN: 1170
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d81b539a32ba7c40a473e6afbb8e0dae525d637&vid=91bf141a7ea82baf5df563f73967884d2a4b5d79&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identification Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR