VWR® AVS TITRINORM Buffer Solution, pH 9 (at 20 °C), Borate free
Supplier: VWR Chemicals
VWRC32037.297EA 119 SGD
VWRC32037.297
VWR® AVS TITRINORM Buffer Solution, pH 9 (at 20 °C), Borate free
Buffers pH Calibration Buffers
Solution prepared from AnalaR® NORMAPUR® grade analytical reagents.
  • Borate free
  • Reference temperature 20 °C
  • True analytical value on the label to an accuracy of ±0,02 pH units
  • Traceable to SRM from NIST
  • Batch number and packing date for traceability
  • Certificate of Analysis available at website
Ordering information: If significantly large volumes are required please contact your local Avantor sales office for a customised offer.