Sodium chloride concentrate for dilution to 0.1 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution

Supplier: VWR Chemicals

Synonyms: Saline, NaCl, Sea salt, Halite

Dose for preparation of 1 litre of solution.

VWRC32064.600EA 62.6 SGD
VWRC32064.600
Sodium chloride concentrate for dilution to 0.1 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution
Sodium chloride
Formula: NaCl
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3

Order Now

Specification Test Results

Titer (20°C) 0.0995 - 0.1005 mol/l


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR