VWR® Boiling Stones ('Anti-Bumping'), Granules
Supplier: VWR Chemicals
VWRC330093YEA 24 SGD
VWRC330093Y
VWR® Boiling Stones ('Anti-Bumping'), Granules
Boiling Stones
Granules of fused alumina, to reduce 'bumping' in boiling liquids.
  • Particle size (1-4 mm): min.96%