VWR® Glass Balls
Supplier: VWR Chemicals
VWRC332124GEA 109 SGD
VWRC332124G
VWR® Glass Balls
Beads Glass Beads
Chemically resistant glass balls suitable for use as mixing beads, boiling stones, or packing for distillation columns.
Order Now
SPECIFICATIONS
Environmentally Preferable