Sodium chloride 99,5, Gen-Apex Molecular_Biology

Supplier: VWR Chemicals

Synonyms: Saline, NaCl, Sea salt, Halite

VWRC33614.265EA 24.1 SGD
VWRC33614.265
Sodium chloride 99,5, Gen-Apex Molecular_Biology
Sodium chloride
Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1413
Melting Pt: 801
Density: 2.16 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599

Order Now

Specification Test Results

Assay Min. 99.5 %
Colouration (1 mol/l; water) Max. 10 APHA
Heavy metals (as Pb) Max. 2 ppm
Transmittance (230 nm) Min. 90 %
Transmittance (260 nm) Min. 95 %
Transmittance (280 nm) Min. 97 %
Transmittance (320 nm) Min. 99 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR