Sodium dodecyl sulphate (SDS) ≥98.5%, Gen-Apex for molecular biology

Supplier: VWR Chemicals
Danger

Synonyms: Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate

VWRC33629.266EA 548 SGD
VWRC33629.266
Sodium dodecyl sulphate (SDS) ≥98.5%, Gen-Apex for molecular biology
Sodium dodecyl sulphate (SDS)
Formula: CH₃(CH₂)₁₁OSO₃Na
MW: 288.38 g/mol
Melting Pt: 204…207 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: 100 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
EINECS: 205-788-1
UN: 1325
ADR: 4.1 III
REACH: 01-2119489461-32
Merck Index: 14,08636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (calculated on anhydrous) Min. 98.5 %
Colouration (0,2 mol/l; water) Max. 10 APHA
Heavy metals (as Pb) Max. 10 ppm
Transmittance (230 nm) (3%) Min. 75%
Transmittance (260 nm) (3%) Min. 80%
Transmittance (280 nm) (3%) Min. 90%
Transmittance (320 nm) (3%) Min. 95%

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR